Learn More
Sino Biological Recombinant Mouse APRIL/TNFSF13 Protein (Trimer, hFc Tag)
Supplier:  Sino Biological 50965M01H11
Encompass_Preferred
A DNA sequence encoding the Mouse TNFSF13 (Q9D777) (Lys104-Leu241) was fused with the Fc region of human IgG1 at the N-terminus. This construct has a trimer design.
Catalog No. 50-299-3605