Learn More
Vector Laboratories Mouse TNFSF13 / APRIL ELISA Kit (Sandwich ELISA)
SDP
Supplier:  Vector Laboratories LSF127941
Encompass
LS-F12794 is a 96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Mouse TNFSF13 / APRIL in samples of Plasma and Serum. It is based upon a Sandwich assay principle and can be used to detect levels of TNFSF13 / APRIL as low as 0.625 nanograms per millilter.
Catalog No. NC1343363