Learn More
Raybiotech Inc Mouse OX40 Ligand (TNFSF4) ELISA Kit
Supplier:  Raybiotech Inc ELMTNFSF41
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 30 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects mouse TNFSF4/OX40 Ligand and on cross reactivity with rmBAFF, rmEDA, rmGITR Ligand, rmFas Ligand, rmLIGHT, rmTWEAK, rhAPRIL, or rhTNFSF4.
Catalog No. 50-195-0147