Learn More
American Research Products Inc Mouse Transforming Growth fact
Supplier:  American Research Products Inc CSBE12862M
Encompass
Mouse Transforming Growth factor b3 TGF-b3 ELISA kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29951