Learn More
World Precision Instrument Mouse Ultra Precise Stereotaxic Instrument
SDP
Supplier:  World Precision Instrument 505316
Encompass
Our Smallest and Lowest Priced Stereotaxic Instrument!
The WPI Mouse Stereotaxic Instruments was specifically designed for use with knock-out and transgenic mice. It is our smallest and most economic stereotaxic instrument. The WPI Mouse Stereotaxic Instruments includes a 100 micron, 3-axes left-hand manipulator arm, mouse snout clamp and tooth bar, dual sided ear bars, and a corner clamp probe holder. Available as Dual, Ultra-Precise, Digital or Motorized.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
Catalog No. 50-253-9128