Learn More
American Research Products Inc Mouse uPA ELISA Kit
Supplier: American Research Products Inc RDRUPAMU
This Mouse Plasminogen Activator Urokinase (uPA) ELISA Kit is a precoated Sandwich ELISA assay designed to detect and quantify the target analyte in serum plasma and other biological fluids This product is for Research Use Only