Learn More
ACROBiosystems ACROBiosystems Mouse VEGF R1 / Flt-1 Protein, Mouse IgG2a Fc Tag, low endotoxin
Supplier:  ACROBiosystems VE1M52561MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (VE1-M5256) is expressed from human 293 cells (HEK293). It contains AA Tyr 23 - Glu 759 (Accession # P35969-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 109.8 kDa. The protein migrates as 140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Mouse VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-M5256) with a linear range of 2-31 ng/mL (QC tested).;Immobilized Human VEGF-B, His Tag (Cat. No. VE6-H5225) at 2 μg/mL (100 μL/well) can bind Mouse VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-M5256) with a linear range of 8-63 ng/mL (Routinely tested).
Catalog No. 50-210-6733