Learn More
Cayman Chemical MousentI6HISIgGSureLIght1ea
SDP
Supplier:  Cayman Chemical 112731 EA
Encompass_Preferred
Mouse anti-6His (6x Histidine) IgG (Clone AD1.1.10) conjugated to SureLight PerCP Antibody: Mouse monoclonal anti-6xHistidine IgG, clone AD1.1.10 Dye: SureLight Peridinin-Chlorophyll (PerCP) Excitation max. λ: 483 nm Emission max. λ: 673 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, Protein microarrays
Catalog No. 50-295-1433