Learn More
Medchemexpress LLC Moxifloxacin | 151096-09-2 | 99.7% | C21H24FN3O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY66011AR100MG
Encompass_Preferred
Moxifloxacin (Standard) is an analytical standard and an orally active 8-methoxyquinolone antimicrobial. It is intended for research and analytical applications, and commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for moxifloxacin.
  • Antimicrobial agent.
  • High purity (99.7%).
  • Used in HPLC, GC, and MS research.
  • Applicable to bacterial sinusitis, chronic bronchitis, and community-acquired pneumonia research.
Catalog No. 50-004-34692