Learn More
Medchemexpress LLC Moxifloxacin | 151096-09-2 | 99.8% | C21H24FN3O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY66011A500MG
Encompass_Preferred
Moxifloxacin is an orally active 8-methoxyquinolone antimicrobial used for treating acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia. It demonstrates rapid action compared to Amoxicillin, showing effects within 3 hours and achieving complete broth sterilization within 24 hours, while also offering protection against macrophage lysis.
  • Orally active 8-methoxyquinolone antimicrobial
  • Treats acute bacterial sinusitis
  • Treats acute bacterial exacerbations of chronic bronchitis
  • Treats community-acquired pneumonia
  • Rapid action compared to Amoxicillin
  • Achieves complete broth sterilization within 24 hours
  • Protects against macrophage lysis
Catalog No. 50-004-33925