Learn More
Medchemexpress LLC Moxifloxacin | 151096-09-2 | MFCD04117996 | 99.7% | 401.43 | C21H24FN3O4 | 10 MG

Supplier: Medchemexpress LLC HY66011AR10MG
Moxifloxacin (Standard) is an analytical reference standard of the fluoroquinolone antibiotic moxifloxacin (free base) provided for research and analytical and quality control applications. The material is characterized for identity and purity and is intended for assay, calibration, method development, and reference testing.
- Purity 99.7% (rounded from supplier listed value).
- Cas number 151096-09-2.
- Molecular formula C21H24FN3O4; molecular weight 401.43.
- Intended for analytical and research use as a reference standard.
- Available in small packaged amounts, for example 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.