Learn More
Apexbio Technology LLC Moxifloxacin(Synonyms: Avelox, Vigamox, Avalox, Moxeza, Proflox, Octegra, MXF, BAY 12-8039), 50mg, CAS: 151096-09-2.
SDP
Supplier:  Apexbio Technology LLC B121850
Encompass_Preferred
Moxifloxacin (CAS 151096-09-2) is a fluoroquinolone antibiotic targeting bacterial DNA gyrase an enzyme essential for DNA replication and transcription It demonstrates broad-spectrum antibacterial activity through inhibition of this enzyme Research indicates that Moxifloxacin exerts dose-dependent antiproliferative and cytotoxic effects in mammalian cell models with concentrations between 50 and 1500 g/mL significantly impacting cell viability and inducing morphological damage such as binucleation Animal studies show intravenous administration (100 mg/kg rats) elevates blood glucose and serum adrenaline levels accompanied by enhanced histamine release but no effect was noted at 75 mg/kg Therefore Moxifloxacin is valuable in biomedical studies exploring antibiotic toxicity cellular proliferation and metabolic responses
Catalog No. 50-000-60406