Learn More
Medchemexpress LLC Moxifloxacin hydrochloride | 186826-86-8 | 99.99% | 1 G
SDP
Supplier:  Medchemexpress LLC HY660111G
Encompass_Preferred
Moxifloxacin hydrochloride (BAY 12-8039) is an oral 8-methoxyquinolone antimicrobial. It is used in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia. This product is for research use only.
  • Oral 8-methoxyquinolone antimicrobial
  • Used for treating various bacterial infections
  • Target: Quinolone
  • For research use only
Catalog No. 50-004-34336