Learn More
Medchemexpress LLC Moxifloxacin hydrochloride monohydrate | 192927-63-2 | MFCD09836237 | 98.0% | 455.91 g/mol | C21H27ClFN3O5 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW353258250MG
Encompass_Preferred
Moxifloxacin hydrochloride monohydrate is the monohydrate salt form of the fluoroquinolone antibiotic moxifloxacin, provided as a research-grade reference for laboratory and analytical applications. It is an orally active antibacterial agent commonly used in studies of respiratory infections and tuberculosis research.
  • Orally active fluoroquinolone antibiotic
  • Intended for research and analytical applications
  • Available in multiple pack sizes, including 250 mg
  • COA and SDS available for traceability and safety
  • Identified by CAS 192927-63-2; formula C21H27ClFN3O5; MW 455.91 g/mol
Catalog No. 50-002-85217