Learn More
Medchemexpress LLC Moxifloxacin (Standard) | 151096-09-2 | 99.7% | C21H24FN3O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY66011AR
Encompass_Preferred
Moxifloxacin is an analytical standard for research and analytical applications. This orally active 8-methoxyquinolone antimicrobial is utilized in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
  • Intended for research and analytical applications
  • Analytical standard for assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-57856