Learn More
Medchemexpress LLC Moxifloxacin (Standard) | 151096-09-2 | 99.7% | C21H24FN3O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY66011AR5MG
Encompass_Preferred
Moxifloxacin (Standard) is the analytical standard of Moxifloxacin, an orally active 8-methoxyquinolone antimicrobial. It is used in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia. This product is for research and analytical applications.
  • Analytical standard for assays
  • Reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Commonly used with HPLC, GC, and MS
Catalog No. 50-004-34285