Learn More
Selleck Chemical LLC Mozavaptan E4957-100MG
SDP
Supplier:  Selleck Chemical LLC E4957100MG
Encompass_Preferred
Mozavaptan hydrochloride(OPC 31260 HCl) a benzazepine derivative is a potent selective competitive and orally active antagonist of vasopressin V2 receptor with an IC50 value of 14 nM Mozavaptan hydrochloride shows 85-fold selectivity for V2 receptor over V1 receptor Mozavaptan hydrochloride has the potential for hyponatremia syndrome of inappropriate antidiuretic hormone (SIADH) and congestive heart failure treatment
Catalog No. 50-000-41788