Learn More
Cayman Chemical mP-2mP-9 InhIbItr I 5mg
SDP
Supplier:  Cayman Chemical 203155 MG
Encompass_Preferred
A potent inhibitor of MMP-2 and MMP-9 (IC50s = 310 and 240 nM, respectively) that is active in vivo as well as in vitro
Catalog No. 50-298-2949