Learn More
Cayman Chemical mP-2mP-9 InhIbItr II 500ug
SDP
Supplier:  Cayman Chemical 24314500 UG
Encompass_Preferred
A dual inhibitor of MMP-2 and MMP-9 (IC50s = 17 and 30 nM, respectively); suppresses lung colonization of Lewis lung carcinoma cells and inhibits tumor-induced angiogenesis, tumor growth, and liver metastasis in mice
Catalog No. 50-293-5013