Learn More
Cayman Chemical mP-8 InhIbItr I 1mg
SDP
Supplier:  Cayman Chemical 218521 MG
Encompass_Preferred
A selective inhibitor of MMP-8 (IC50 = 4 nM); does not target other MMPs
Catalog No. 50-296-4055