Learn More
Cayman Chemical mP-9mP-13 InhIbItr I 1mg
SDP
Supplier:  Cayman Chemical 212651 MG
Encompass_Preferred
A cell-permeable inhibitor of MMPs that most potently inhibits MMP-9 and MMP-13 (IC50s = 0.9 nM for both); less effectively inhibits MMP-1, MMP-3, and MMP-7 (IC50s = 43, 23, and 920 nM, respectively)
Catalog No. 50-298-1456