Learn More
Cayman Chemical mP InhIbItr II 10mg
SDP
Supplier:  Cayman Chemical 1595810 MG
Encompass_Preferred
A reversible, broad-range inhibitor of MMP-1 (IC50 = 24 nM), MMP-3 (IC50 = 18.4 nM), MMP-7 (IC50 = 30 nM), and MMP-9 (IC50 = 2.7 nM)
Catalog No. 50-297-2076