Learn More
Cayman Chemical mP InhIbItr ItrIfluoROC 500ug
SDP
Supplier:  Cayman Chemical 24708500 UG
Encompass_Preferred
An inhibitor of MMP-1, MMP-2, and MMP-3 (IC50s = 1.3, 30, and 150 μM, respectively); selective for MMP-1, MMP-2, and MMP-3 over the proteases thermolysin, urease, trypsin, α-chymotrypsin, plasmin, and elastase (4 mM)
Catalog No. 50-293-6689