Learn More
Chemglass Life Sciences MPC to MPX Back-to-Back Insert Adapter, CPC MPC22X2239M
SDP
Supplier:  Chemglass Life Sciences CLS9141303X
Encompass
MPC Series couplings add ease of use and security to your most critical fluid handling applications. Choose from a full line of connectors and configurations, including pressure sealing caps and plugs, in sizes to fit 1/8", 1/4" and 3/8" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks. Features: Ergonomic thumb latch USP Class VI materials Sterilizable by autoclave, Et0, e-beam or gamma Parting line-free hose barb BSE/TSE free materials Benefits: Easy to operate - even with gloved hands Meet biocompatibility requirements Reusable, yet economical enough to allow disposability Eliminates potential leak path Meet ADCF requirements.
Catalog No. 50-009-80631