Learn More
Medchemexpress LLC mPEG-CM MW 2000 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW1048558A
Encompass_Preferred
mPEG-CM (MW 2000) (mPEG-COOH (MW 2000)) has a reactive carboxyl group (-COOH) attached to the terminal site of the structure which can form a stable amide bond with the amino group or an ester bond with the hydroxyl group mPEG-CM (MW 2000) can be used for drug delivery[1]
Catalog No. 50-002-04606