Learn More
Medchemexpress LLC mPEG-N3 MW 2000 25mg
SDP
Supplier:  Medchemexpress LLC 25MGHYW1049092
Encompass_Preferred
mPEG-N3 (MW 2000) can be used to modify proteins peptides and other materials The azide group can react with alkynes in aqueous solution catalyzed by copper It can also be easily reduced to an amine group PEGylation can increase solubility and stability and reduce the immunogenicity of peptides and proteins mPEG-N3 (MW 2000) can also inhibit the nonspecific binding of charged molecules to the modified surface[1]
Catalog No. 50-002-04686