Learn More
Creative Pegworks mPEG-SCM, MW 2000, 1 g
SDP
Supplier:  Creative Pegworks PLS2141G
Encompass
mPEG-SCM is one type of mPEG-NHS reagents. There is a methylene (CH2) linkage between PEG and the NHS ester.
Catalog No. NC1528163