Learn More
Medchemexpress LLC MPEP | 96206-92-7 | 99.4% | 193.24 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14609A100MG
Encompass_Preferred
MPEP is a potent, selective, noncompetitive, orally active, and systemically active mGlu5 receptor antagonist. It completely inhibits quisqualate-stimulated phosphoinositide (PI) hydrolysis with an IC50 of 36 nM. MPEP exhibits anxiolytic- and antidepressant-like effects. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent, selective mGlu5 receptor antagonist
  • Exhibits anxiolytic-like effects
  • Shows antidepressant-like effects
  • Functions as a click chemistry reagent
  • No agonist or antagonist activity on human mGlu2, -3, -4a, -7b, -8a, and mGlu6 receptors
Catalog No. 50-003-60095