Learn More
Cayman Chemical MPEPhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1453610 MG
Encompass_Preferred
A potent, highly selective, and centrally active antagonist at the mGlu5a receptor subtype (IC50 = 36 nM); also has been reported as a positive allosteric modulator of mGluR4 at higher concentrations
Catalog No. 50-296-9391