Learn More
Medchemexpress LLC Mpg peptides, Pβ | 791642-10-9 | 98.3% | 2807.32 | 5 MG
Supplier: Medchemexpress LLC HYP40765MG
Mpg peptides, Pβ is a primary amphiphilic peptide consisting of three domains. It is an amphipathic cell-penetrating peptide that forms stable non-covalent complexes with nucleic acids (including DNA) through electrostatic interactions, enhancing their intracellular delivery. It can be used in studies of HIV-1-related immune responses.
- Consists of three domains
- Forms stable non-covalent complexes with nucleic acids
- Enhances intracellular delivery of nucleic acids
- Useful in HIV-1 related immune response studies