Learn More
Medchemexpress LLC mPGES1-IN-7 | 1892595-16-2 | 95.5% | 347.37 g/mol | C15H14FN5O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11736425MG
Encompass_Preferred
HS148 is a small-molecule research compound that selectively inhibits death-associated protein kinase 3 (DAPK3), with a reported Ki of 119 nM. Supplied as a dry powder, it has a listed purity of 95.5% and a molecular weight of 347.37 g/mol. Intended for biochemical and cellular research use only.
  • Selective DAPK3 inhibitor with a reported Ki of 119 nM.
  • Listed purity of 95.5%, suitable for research applications.
  • Molecular weight 347.37 g/mol for characterization and dosing.
  • Supplied as a stable dry powder for convenient storage and handling.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-28764