Learn More
Selleck Chemical LLC MPI-0479605
SDP
Supplier:  Selleck Chemical LLC S74881MG
Encompass_Preferred
MPI-0479605 is an ATP competitive and selective inhibitor of mitotic kinase Mps1 with IC50 of 1 8 nM 40-fold selectivity over other kinases
Catalog No. 50-000-02864