Learn More
Genetex Inc Myeloperoxidase Monoclonal Antibody [MSVA-692M] HistoMAX™ (GTX04470) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04470
Encompass_Preferred
The Myeloperoxidase antibody [MSVA-692M] HistoMAX™ (GTX04470) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human spleen tissue using GTX04470 Myeloperoxidase antibody [MSVA-692M] HistoMAX. Numerous myeloperoxidase positive granulocytes are usually seen within the red pulp of the spleen.
Catalog No. NC3667761