Learn More
Cayman Chemical MPO asy Buffer1X 1ea
SDP
Supplier:  Cayman Chemical 7001641 EA
Encompass_Preferred
A - and K1-opioid receptor antagonist (Kis 0.3 and 0.2 nM respectively) selective for - and K1-opioid receptors over the -opioid receptor (Ki 12.8 nM) reversibly inhibits electricity-induced twitches of isolated guinea pig ileal longitudinal muscle strips (IC50 48 nM) indicating opioid agonist activity inhibits endomorphin 1-induced conditioned place preference in mice at 0.3 UG/animal i.c.v. prevents decreases in urine output induced by fentanyl D-propoxyphene buprenorphine profadol or bromadoline in water-loaded rats at 40 mg/kg reduces hemokinin 1-induced increases in the latency to withdrawal in the tail-flick test in mice
Catalog No. 50-289-3703