Learn More
Medchemexpress LLC MPO-IN-3 | 1435469-45-6 | 99.2% | 371.88 g/mol | C16H22ClN3O3S | 5 MG

Supplier: Medchemexpress LLC HY1451975MG
MPO-IN-3 is a potent myeloperoxidase (MPO) inhibitor described in WO2013068875A1 (example 191). It is provided as a research compound for in vitro and biochemical studies of MPO activity, with a CAS registry number and high reported purity suitable for laboratory use.
- Potent myeloperoxidase inhibitor suitable for MPO activity studies.
- High reported purity (99.17%) for reproducible results.
- Molecular weight 371.88 g/mol and formula C16H22ClN3O3S.
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance may be required.
- Recommended storage under nitrogen at 4°C; in solution store at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.