Learn More
Hycult Biotech Inc MPO MOUSE ELISA KIT 2X96 TESTS
SDP
Supplier:  Hycult Biotech Inc HK21002
Encompass
The mouse MPO ELISA has been developed for the quantitative measurement of natural and recombinant mouse MPO in cell culture medium, plasma and tissue homogenates.
Catalog No. NC0045704