Learn More
GT MOLECULAR LLC MPOX ASSAY KIT FOR QIACUITY
SDP
Supplier:  GT MOLECULAR LLC 1004591.1
Encompass
  • This assay kit enables the detection and quantification of non-variola Orthopoxviruses and Mpox Clade II in wastewater samples.
  • Designed for seamless integration with the QIAGEN QIAcuity Digital PCR System.
  • Complete with all necessary primers, probes, and controls.
  • Facilitates streamlined workflows for differentiating between multiple pathogens.
  • Intended for research use only, not for diagnostic procedures.
  • Optimized design leads to time, cost, and reagent savings.
  • Ultrasensitive detection capabilities ensuring high output efficiency.
  • Recognizes both Clade 1 mpox (including Clade 1b) and Clade 2 mpox.
  • Supports environmental surveillance efforts effectively.
  • Limited to 200 reactions per kit.
Catalog No. NC3051615