Learn More
Cayman Chemical MPP+ Iodide | 36913-39-0 | MFCD00055096 | 100mg
SDP
Supplier:  Cayman Chemical 16958100
Encompass_Preferred
MPP+ Iodide | Purity: ≥98% | MW: 297.1 | 36913-39-0 | MFCD00055096 | 100mg
Catalog No. NC1759057