Learn More
Medchemexpress LLC Mpro inhibitor N3 | 884650-98-0 | 99.8% | 680.79 g·mol⁻¹ | C35H48N6O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1361495MG
Encompass_Preferred
Mpro inhibitor N3 is a peptide-based inhibitor of the SARS-CoV-2 main protease (MPro) used as a biochemical and antiviral research reagent. It has a reported EC50 of 16.77 μM against SARS-CoV-2 and shows antiviral activity versus HCoV-229E, FIPV, IBV, and MHV-A59; molecular formula C35H48N6O8 and molecular weight 680.79 g·mol⁻¹.
  • Inhibits SARS-CoV-2 main protease (MPro).
  • Reported EC50 of 16.77 μM against SARS-CoV-2.
  • Demonstrates antiviral activity against multiple coronaviruses.
  • High reported purity (99.76%).
  • Suitable for biochemical assays and antiviral research.
  • Available in small-mass research quantities for laboratory use.
Catalog No. 50-002-22490