Learn More
Medchemexpress LLC MPSD control peptide (TFA) | 95.8% | 3192.80 | 1 MG

Supplier: Medchemexpress LLC HYP10471E1MG
MPSD control peptide (MARCKS-ED control peptide) is a control peptide for MPSD peptide (HY-P10471). This 25-amino acid peptide is based on the effector domain sequence of the intracellular membrane protein myristoylated alanine-rich C-kinase substrate (MARCKS). It can sense membrane curvature and recognize phosphatidylserine, making it useful as a biological probe to study membrane shape and lipid composition.
- Control peptide for MPSD peptide
- Composed of 25 amino acids
- Based on MARCKS effector domain sequence
- Senses membrane curvature
- Recognizes phosphatidylserine
- Utilized as a biological probe
- Formula: C151H243N41O35.xC2HF3O2
- Appearance: White to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.