Learn More
Selleck Chemical LLC MPTP hydrochloride S4732-100mg
SDP
Supplier:  Selleck Chemical LLC S4732100MG
Encompass_Preferred
MPTP hydrochloride is a dopaminergic neurotoxin and cause selective destruction of dopaminergic neurons in animal models of parkinsonism MPTP hydrochloride induces apoptosis MPTP hydrochloride can be used to induce animal models of Parkinson s disease
Catalog No. 50-311-1975