Learn More
Sino Biological Monkeypox Virus (MPXV) (Clade Ib) Envelope protein E8L Protein (His Tag)
Supplier:  Sino Biological 41042V08H100
Encompass_Preferred
A DNA sequence encoding the Monkeypox Virus (MPXV) E8L Protein (EPI_ISL_19093796) (Met1-Thr275) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3324