Learn More
Medchemexpress LLC Mr-l2 | 2374703-19-0 | 98.6% | 441.71 g/mol | C19H16Cl3FN4O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1283585MG
Encompass_Preferred
MR-L2 is a reversible, noncompetitive allosteric activator of long-isoform phosphodiesterase-4 (PDE4) used as a research tool in biochemical and pharmacological studies. It is provided as a high-purity small-molecule research compound with common solubility in DMSO and is intended for in vitro assay and mechanism-of-action work.
  • Activates PDE4 long isoforms via an allosteric, noncompetitive mechanism.
  • High reported purity for reliable assay results.
  • Available as solid and as DMSO solution formats for assay flexibility.
  • Supplied in small milligram quantities suitable for biochemical testing.
  • Molecular formula C19H16Cl3FN4O; molecular weight 441.71 g/mol for reference.
Catalog No. 50-002-64411