Learn More
Medchemexpress LLC MR-L2 | 2374703-19-0 | 98.6% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12835850MG
Encompass_Preferred
MR-L2 is a reversible and noncompetitive allosteric activator of long-isoform phosphodiesterase-4 (PDE4). It activates representative PDE4 long-isoform variants including PDE4A4, PDE4B1, PDE4C3, and PDE4D5. This compound suppresses PGE2-induced MDCK cell cyst formation with an EC50 of 1.2 μM and significantly suppresses cAMP elevation and cysts formation in MDCK cells. It is intended for research use only.
  • Reversible and noncompetitive allosteric activator of long-isoform PDE4
  • Activates PDE4A4, PDE4B1, PDE4C3, PDE4D5 variants
  • Suppresses PGE2-induced MDCK cell cyst formation
  • Suppresses cAMP elevation in MDCK cells
Catalog No. 50-003-41955