Learn More
Apexbio Technology LLC MRE 3008F20 252979-43-4 10mg
SDP
Supplier:  Apexbio Technology LLC B551810
Encompass_Preferred
MRE 3008F20 is a small-molecule antagonist targeting the TRAIL receptor signaling pathway It is designed to inhibit activation of the TRAIL receptor thereby blocking tumor necrosis factor-related apoptotic processes MRE 3008F20 exerts its biological activity by specifically binding to the ligand-binding site of the TRAIL receptor (TRAIL-R) preventing receptor activation and inhibiting downstream apoptosis signaling events Based on these pharmacological properties MRE 3008F20 holds research potential in cell biology assays oncology pharmacology studies and investigations using disease-relevant cellular and animal models
Catalog No. 50-000-88729