Learn More
TARGETMOL CHEMICALS INC MRE3008F20 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T161325MG
Encompass_Preferred
Also available in 1 mg, 2 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. MRE3008F20 is a species-selective and potent adenosine A3 receptor (AA3R) antagonist that inhibits Cl-IB-MECA-induced cAMP production and can be used in glaucoma and asthma studies.13483-88-88-9 Purity 97.66%
Catalog No. 50-287-6074