Learn More
Cayman Chemical MreBPRturbngCompoundA2225mg
SDP
Supplier:  Cayman Chemical 1587025 MG
Encompass_Preferred
A benzylisothiourea compound that interacts with the ATP binding site of MreB rapidly and reversibly; blocks normal rod shape formation and inhibits chromosome partitioning in E. coli, inhibiting growth (MIC = 3.1 UG/ml); interferes with chromosome segregation in C. crescentus
Catalog No. 50-296-9458