Learn More
Medchemexpress LLC MRGPRX1 agonist 1 | 2377379-39-8 | 99.95% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13011810MM/1ML
Encompass_Preferred
MRGPRX1 agonist 1 is a highly potent and selective agonist for MRGPRX1, with an EC50 of 50 nM. It exhibits over 50-fold selectivity compared to δ, μ, and κ opioid receptors and is inactive against MRGPRC11. This compound is valuable for studying chronic pain, particularly neuropathic pain.
  • Highly potent MRGPRX1 agonist
  • Greater than 50-fold selectivity for δ, μ, and κ opioid receptors
  • Inactive against MRGPRC11
  • Useful for the study of chronic pain
  • EC50 of 50 nM for MRGPRX1
  • Inhibits high voltage-activated (HVA) Ca2+ currents
  • Reduces neurotransmitter release
  • Mitigates nociceptive transmission in the spinal cord
  • Shows good distribution to the spinal cord in male mice
Catalog No. 50-003-39014