Learn More
Medchemexpress LLC MRIA9 | 2750707-05-0 | 98.9% | 496.92 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY139253
Encompass_Preferred
MRIA9 is a chemical probe that acts as an ATP-competitive, pan Salt-Inducible kinase (SIK) and PAK2/3 inhibitor. It has IC50 values of 516 nM for SIK1, 180 nM for SIK2, and 127 nM for SIK3.
  • ATP-competitive inhibitor
  • Pan Salt-Inducible kinase (SIK) inhibitor
  • PAK2/3 inhibitor
  • High oral bioavailability (F = 75-80%)
Catalog No. 50-003-49184