Learn More
Enzo Life Sciences MRP4 monoclonal antibody (M4I-10) (75µg)
SDP
Supplier:  Enzo Life Sciences ALX801038C075
Encompass_Preferred
Clone: M4I-10. Isotype: IgG2a. Immunogen: Recombinant human MRP4 (multidrug resistance-associated protein 4) (aa 372-431). Species reactivity: Human, Mouse. Applications: ICC, IHC (FS), WB. Formulation: Liquid. In serum free culture supernatant containing 0.7% BSA and 0.1% sodium azide.
Catalog No. 50-200-9708